Learn More
Cytiva Whatman™ Clear Polystyrene Universal Lid
Lids are suitable for using as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory.
Brand: Cytiva 7704-1001
179.27 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Specifications
Clear | |
100 Pieces | |
Universal Lid |
Polystyrene | |
Lids are suitable for using as dust covers and to prevent splashing or contamination when plates are being moved around the laboratory. |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now