missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid
Whatman™ (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.
191.00€ - 201.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Bubble Point | 1.3 bar |
Color | White |
Material | MCE |
Material (Membrane) | ME 27 |
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | Price | Quantity & Availability | |||||
11909227
|
Cytiva
10409970 |
Gridded with Pad |
201.00€
Pack of 100 |
Estimated Shipment: 14-03-2025
Log in to see stock. |
|||||
11959217
|
Cytiva
10408970 |
Gridded without Pad |
191.00€
Pack of 100 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 27
- Pore Size: 0.8 μm
- Size (Metric) Grid: 3.1 mm
- Color: White
- Grid Color: Black
- Autoclavable: Yes
- Sterility: Sterile
- Thickness (Metric): 140 μm
- Diameter (Metric): 47 mm
- Flow Rate: 2.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
- Bubble Point: 1.3 bar
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
White | |
ME 27 | |
Sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Black | |
3.1 mm | |
140 μm | |
47 mm |
1.3 bar | |
MCE | |
0.8 μm | |
2.8 sec./100 mL/12.5 cm ² (at Δp = 0.9 bar) | |
11.6 sec./100 mL (at Δp = 3 mbar) | |
100 Pk. | |
0.006 in. | |
1.85 in. | |
47 mm |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose