missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid
Whatman™ (ME 27) Mixed Cellulose Ester Membranes, White/Black Grid are particularly effective in applications requiring higher flow rates and larger volume filtration. Supplied with clearly defined grid lines at regularly spaced intervals to assist in microbiological colony counting.
Brand: Cytiva 10409970
This item is not returnable.
View return policy
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 27
- Pore Size: 0.8 μm
- Size (Metric) Grid: 3.1 mm
- Color: White
- Grid Color: Black
- Autoclavable: Yes
- Sterility: Sterile
- Thickness (Metric): 140 μm
- Diameter (Metric): 47 mm
- Flow Rate: 2.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
- Bubble Point: 1.3 bar
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
White | |
MCE | |
0.8 μm | |
2.8 sec./100 mL/12.5 cm ² (at Δp = 0.9 bar) | |
11.6 sec./100 mL (at Δp = 3 mbar) | |
100 Pk. | |
0.006 in. | |
1.85 in. | |
47 mm |
1.3 bar | |
Gridded with Pad | |
ME 27 | |
Sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Black | |
3.1 mm | |
140 μm | |
47 mm |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose