Learn More
Invitrogen™ West Nile Virus Envelope Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0222
Description
West Nile Virus Envelope Synthetic Peptide, PEP-0222, from Invitrogen.
West Nile Virus (WNV) is a member of the Flaviviridae, a plus-stranded virus family that includes St. Louis encephalitis virus, yellow fever virus, and Dengue virus. WNV was initially isolated in 1937 in the West Nile region of Uganda and has become prevalent in Africa, Asia, and Europe. It has rapidly spread across the United States with cases being observed in every continental state. Virus particles consist of a dense core made up of the core/capsid protein encapsulating the RNA genome surrounded by a membrane envelope embedded with envelope and matrix proteins which play a major role for WNV entry into target cells. The viral core protein is thought to contribute to the WNV-associated inflammation via apoptosis induced though the caspase-9 pathway as delivery of core gene delivery into the striatum of mouse brain and skeletal muscle resulted in cell death and inflammation.
Specifications
Virus | |
West Nile Virus Envelope | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
0.02% sodium azide | |
50 μg | |
PBS |
purified | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
No | |
Peptide | |
RUO - research use only |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.