Learn More
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™ Product Code.: 10327873
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™
Product Code. 10327873
1 L
Click to view available options
Quantity:
1 L
2.5 L
Product Code. 10327873
Brand: Fisher Chemical W/0110/PB15
Request Bulk or Custom Format
For use with HPLC gradient analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Plastic Coated Glass Bottle
Quantity 1 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527436 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2526366 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2525710 Certificate TypeCertificate of Analysis Date06/02/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Product Identifier
  • Water
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
Product Content Correction
Product Title
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™ >