missing translation for 'onlineSavingsMsg'
Learn More

Rogo Sampaic™ Watch Glass Dishes Ground Edge, 70 mm

Concave watch glasses made of glass pieces that present ground edges.

Brand:  Rogo Sampaic™ KBN004

Product Code. 11758094

  • 46.00€ / Pack of 10
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These Endo Glassware watch glasses are concave, made of glass pieces that present ground edges.

Can be used as:

  • Weighing trays
  • For drying wet solids
  • For the observation of small invertebrates under a stereomicroscope
  • As recipients in oil tasting
  • As a lid for beakers, flasks and other recipients
Specifications

Specifications

Glass
To Evaporate Liquids or Weigh Samples
2.78 in.
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rogo Sampaic™ Watch Glass Dishes Ground Edge, 70 mm > Watch Glass; Material: Glass; Diameter (Metric): 70 mm; For Use With (Application): To Evaporate Liquids or Weigh Samples

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.