Learn More
Haleco™ Waste Disposal Bags
226.00€ - 241.00€
Specifications
Product Type Garbage Bag
Color Gray
Disposable Single-use
For Use With (Application) Multi-purpose
Material LDPE
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
12536239
missing translation for 'viewDocuments'
Haleco™
SACG100ST
100 L
226.00€
Pack of 200
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
12526239
missing translation for 'viewDocuments'
Haleco™
SACG50ST
50 L
241.00€
Pack of 500
Estimated Shipment: 15-05-2025
to see stock.
Specifications
Garbage Bag
Single-use
LDPE
Gray
Multi-purpose
Product Title
Haleco™ Waste Disposal Bags