missing translation for 'onlineSavingsMsg'
Learn More

Kinematica HMC Europe Vertical Autoclaves: HG Series

Self contained top loading autoclaves offer large capacity in a compact unit and easy to use controls.

Brand:  Kinematica HMC Europe 21002

 View more versions of this product

Product Code. 10003453

  • 18950.00€ / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Programmable top-loading autoclaves/sterilizers

  • Use steam sterilization
  • Six sizes from 25 to 110 liters
  • Low height, easy-to-load
  • Portable — require no plumbing to operate but can be plumbed
  • Complete line of baskets, inserts and cans available separately
  • HG Series (advanced features)
  • Many upgrades available depending on your needs

Certifications

CE marked

Services

Two years on parts; One year on labor

Specifications

Specifications

CE
69.1 cm
80 L
36.4 cm
40.55 in.
HG 80
105°C to 135°C
73 kg
28.74 in.
3.8 w
45.5 cm
27.2 in.
21.13 gal.
14.33 in.
Steam sterilization
103 cm
221°F to 275°F
160.93 lb.
73 cm
2.5 psi
17.91 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions