Learn More
VACUUBRAND™ VARIO™ Chemistry Pumping Unit, PC 3012 VARIO Select
Provides pinpoint vacuum control by adjusting the motor speed. This model supports high pumping speeds and includes an exhaust vapor condenser. Suitable for use with floor-standing rotary evaporators, vacuum drying ovens, and work in miniplants, kilo labs, and scale-up facilities.
Brand: VACUUBRAND™ 20743875
This item is not returnable.
View return policy
Description
- VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
- Automatic boiling point detection and adjustment of the vacuum for short process times
- Meets high vacuum requirements for large quantities of most high boiling solvents
- Extraordinary diaphragm life for minimum operational and servicing costs
- Excellent environmental compatibility through low energy consumption and efficient solvent recovery
- High pumping speed and deep ultimate vacuum is suitable for demanding applications with high boiling point solvents
- Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
- For solvent evaporation, the VACUU·SELECT controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control
- VARIO motor speed control for exceptionally long maintenance intervals
- Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
- Touch screen display can be operated even when wearing safety gloves
- Choose from 14 languages
Specifications
1.5 Mbar (Without Gas Ballast), 3 Mbar (With Gas Ballast) | |
35 kg | |
30-2400 min-1 | |
616 x 435 x 450 mm | |
PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM | |
47 dBA | |
1 kW | |
10°C | |
50-60 Hz | |
IP 40 | |
Intelligent Vacuum with VARIO™-control, Drying Chambers, Filtration and Solid Phase Extraction, Local Area Vacuum Networks for Laboratories | |
1.1 bar |
VACUU·SELECT | |
Ready-to-Use Pumping Unit (Completely Mounted with Manual) | |
CE-certification; II 3/-G IIC T3 x Internal Atm. Only | |
Peltronic™ Emission Condenser works without any cooling media like water or dry ice | |
PC 3012 VARIO Select EKP | |
Kaltgerätestecker EN 60320 | |
40°C | |
15 m3/hr. | |
Diaphragm Pump | |
8 | |
3 | |
200/230 V |