missing translation for 'onlineSavingsMsg'
Learn More
Vancomycin hydrochloride, MP Biomedicals™ Product Code.: 11485492
compliance-icons

Vancomycin hydrochloride, MP Biomedicals™

Product Code. 11485492 Shop All MP Biomedicals Products
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 11485492

Brand: MP Biomedicals 0219554005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Type Vancomycin Hydrochloride
Physical Form Powder
CAS 123409-00-7
Color White
Quantity 5 g
Molecular Formula C66H257Cl2N9O24
InChI Key ARNOCQCQWLXLRG-UHFFFAOYSA-N
SMILES C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl
IUPAC Name azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride
Molecular Weight (g/mol) 1632.721
PubChem CID 44134997
Formula Weight 1485.72
Chemical Name or Material Vancomycin hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Vancomycin Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vancomycin hydrochloride, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.