Learn More
Thermo Scientific™ Vacuum Hydrolysis Tube
Perform derivatization and other small-scale reactions reliably and with ease.
Brand:  Thermo Scientific™ TS-29570
232.75 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13150093
  • 233.91€ / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
Ideal applications
  • General sample incubation and evaporation in a variety of tube and vial sizes
  • Silylation, alkylation, and acylation derivatization reactions for GC sample preparation
  • Protein hydrolysis and vacuum hydrolysis reactions for amino acid analysis by HPLC
Additional features
  • LED display: match digital display to in-block thermometer to calibrate temperature set-point
  • Modular design: switch aluminum blocks and vials; attach compatible evaporator manifold
  • Four models: single-block and triple-block sizes with either heat-only or heat-and-stir capability
  • In-block temperature control option by Remote Temperature Probe, an optional accessory to allow temperature regulation from block wells or actual sample vials
Complete Reacti-Therm Sample Derivatization System
The derivatization system can combine heating, stirring and evaporation in a versatile, modular system that is easily configured to your application.
  • Reacti-Therm Heating/Stirring Module
  • Reacti-Vap Evaporator Module
  • Reacti-Blocks, Reacti-Vial Magnetic Stirrers, and Reacti-Vial Small Reaction Vials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 mL
1
Documents
One moment while we fetch your results.
Product Title
Thermo Scientific™ Vacuum Hydrolysis Tube > Vacuum Hydroysis Tube, 1 mL