Learn More
Eppendorf™ VacuTip II
Eppendorf™ VacuTip II provides you with the best microcapillary for cell manipulation applications.
Brand:  Eppendorf™ 5195000044
Product Code. 15881535
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • 25° tip angle
  • Flange length 0.5mm
  • Smooth rounded tip end for gentle holding
  • Sterilized via validated gamma irradiation
  • Non-cytotoxic (as proven by mouse embryo assay)
  • Capillary safe for optimum protection
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60 μm
110 μm
Product Title
Eppendorf™ VacuTip II > Diameter Metric Inside: 60μm