missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Z No Additive Blood Collection Tubes

VACUETTE™ Z No Additive Collection Tubes are used in determinations for clinical chemistry and hormones, microbiological serology, immunology, TDM and crossmatching. These white top no additive blood tubes are typically used as discard tubes or as an alternative to serum tubes with a clot activator. They contain no chemical additives, are evacuated and sterile.

Brand:  Greiner Bio-One 454001

 View more versions of this product

Product Code. 15432520

  • 9.17€ / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

VACUETTE™ Z No Additive Collection Tubes are used in determinations for clinical chemistry and hormones, microbiological serology, immunology, TDM and crossmatching. These white top no additive blood tubes are typically used as discard tubes or as an alternative to serum tubes with a clot activator. They contain no chemical additives, are evacuated and sterile.

  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Evacuated and are sterile
  • Made out of virtually unbreakable PET plastic
  • They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Contain no additive in the tube. After one hour the blood in the tube is completely coagulated
  • CE marked
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blood Collection Tube
Sterile
4 mL
VACUETTE™ Blood Collection Needles
75 mm
Blood collection
13 mm
No Additive
Premium Safety Twist Cap
13 mm
White
13 x 75 mm
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.