missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Vacuette™ Glucose Blood Collection Tube FE Sodium Fluoride/K3EDTA

Vacuette™ Blood Collection Tubes have set the standard in today‘s marketplace. They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass.

Brand:  Greiner Bio-One 454085


Product Code. 13078652

  • 421.00€ / Pack of 1200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Made from virtually unbreakable PET plastic
  • Optically clear as glass
  • Contains sodium fluoride as a glycolysis inhibitor
  • Anticoagulant K3EDTA prevents blood from clotting
  • Citrate/citric acid buffer reduces the pH value in the sample immediately after blood collection leading to an instant stabilisation of glucose
  • Improved safety and hygiene during specimen collection procedures
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium Fluoride/K3 EDTA
Sterile
2 mL
13 mm x 75 mm
PET
Screw Top
Gray with White Ring
Blood glucose analysis
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X1200 Blood tube Vacuette Glucose EDTA/NaFl pack of 1,200 tubes grey cap 2mL

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic Acid Tripotassium Dihydrate, Sodium Fluoride

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.