Learn More
Cytiva VacuCap™ 90 Vacuum Filtration Devices
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
Brand: Cytiva 4634
208.05 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
This item is not returnable.
View return policy
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Supor (Hydrophilic PES) | |
Sterile | |
60 mm |
0.45 μm | |
30 cm2 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Promotions
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
47Hours
13Minutes
21Seconds
22New Offer Save Up to 70%
Product Content Correction
Product Title
Cytiva VacuCap™ 90 Vacuum Filtration Devices > 0.45 μm, 60 mm