Learn More
Syngene™ UV Safety Shield
Safety shield designed to offer UV protection.
Brand: Syngene™ UV-SHIELD-LARGE
This item is not returnable.
View return policy
Description
- The wearing of safety equipment according to laboratory regulations is recommended
- Large Shield Dimensions: 14 W x 24 in. H
Specifications
Syngene | |
GBox Chemi XX6 and XX9 | |
Large | |
Safety Shield |
UV Protection | |
UV Safety Shield, 24 in. high | |
110/230 V |
Product Content Correction
Product Title
Syngene™ UV Safety Shield > UV Safety Shield; Size: Large