Learn More
Grant Instruments™ UV Cabinet
UVT-B-AR Economy bench-top model for protection against contamination during a variety of DNA/RNA procedures, with dual UV lamp protection.
Brand:  Grant Instruments™ UVT-B-AR EURO
2225.92 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 13424356
  • 3390.00€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • UV surface irradiation – via single 25W 254nm open UV lamp
  • Patented high intensity UV air cleaner – 25 m3/hour cleaner recirculator continuous air flow with 1cm UV irradiation distance
  • UV exposure control – 24 hour digital timer
  • Built-in power socket
  • UV protection - UV protective film on glass panels
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • White lamp provides local illumination of the workplace to optimise visual control during operations
  • Shock proof glass front, stainless steel sides, metal framework and stainless steel work surface
  • Patented built-in UV cleaner recirculator prevents unwanted contamination and protects the user from direct UV light during manipulation
  • Second UV lamp disinfects the working area, inactivating DNA/RNA fragments during 15-30 min of exposure. Automatic switch-off when door is opened
  • Contains an integral power socket
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UV Cabinet
Stainless Steel, Glass
585 mm
7 m3/h
50/60 Hz
120/230 V
650 mm
1 open UV lamp 25 W bactericidal, 254 nm, ozone free, 1 UV recirculator, 25 W (efficiency >99% per 1 cycle), 1 white lamp
0.3 A
475 mm
555 mm
Bactericidal air recirculator, 25m3/h air flow exchange
690 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ UV Cabinet >