Learn More
Kinesis™ Adapters for HPLC
Convenience of quick connect and disconnect of a luer connection
Brand:  Kinesis™ P-625
 View more versions of this product
Product Code. 15136063
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
Adapter
Male(External) threads Luer to 1/4-28 Flat-bottom
Natural
0.04 in.
ETFE
Male(External) Threads Luer to 1/4-28 Flat-Bottom, Male(External) Threads