Learn More
Kinesis™ Adapters for HPLC
Click to view available options
Connections:
Female Luer to Male 10-32 Adapter
Male Luer to 1/4-28 Flat Bottom
Material:
ETFE
Unit Size:
Each
Description
Convenience of quick connect and disconnect of a luer connection
- Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
| Type | Adapter |
| Diameter (English) Inner | 0.04 in. |
| Connections | Male Luer to 1/4-28 Flat Bottom |
| Material | ETFE |
| Color | Natural |