Learn More
Kartell™ Universal Safety Pipette Bulb
Reduce risk of cracking or chipping when filling any size pipette. Kartell™ Universal Safety Pipette Bulb uses an attchment with an adapter to seal around pipettes without twisting.
Brand: Kartell™ 0020800
9.26 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Reduce risk of cracking or chipping when filling any size pipette. Kartell™ Universal Safety Pipette Bulb uses an attchment with an adapter to seal around pipettes without twisting.
- Red rubber with silicon bulb
- For use with pipettes with a capacity of up to 25mL
Specifications
Rubber, Silicone | |
Universal Pipette Filler |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%