missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UNC 1999

Potent and selective EZH2/EZH1 inhibitor

286.00€ - 1110.00€

Specifications

Purity >99%
Product Type UNC 1999
Molecular Weight (g/mol) 569.74
Inhibitors UNC 1999
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18779528
View Documents Promotion Details
Tocris Bioscience™
4904/10
10 mg
286.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18790929
View Documents
Tocris Bioscience™
4904/50
50 mg
1110.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.

Negative Control also available.

Specifications

Specifications

>99%
UNC 1999
569.74
UNC 1999
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only