Learn More
Tocris Bioscience™ UNC 1999
Potent and selective EZH2/EZH1 inhibitor
Brand:  Tocris Bioscience™ 4904/10
221.16 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18779528
  • 286.00€ / 10mg
Explore more special offers
Description
Description
UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.
Negative Control also available.
Specifications
Specifications
>99%
UNC 1999
UNC 1999
10 mg
569.74