Learn More
IKA™ ULTRA-TURRAX™ T 18 Digital Disperser
GREENER_CHOICE
Disperses volumes from 1 to 1500mL (H2O) with digital speed display. IKA™ ULTRA-TURRAX™ T 18 Digital Disperser offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
Brand:  IKA™ 0009022800
2195.76 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 13470657
  • 2638.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Competitively priced dispersing instrument for volumes from 1 to 1500mL (H2O) with digital speed display. Offers a wide speed range from 3000 to 25,000 rpm that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
  • Motor protection against overload
  • Digital display for precise monitoring of set and actual speeds 2+1 year after registering at 2+1 www.ika.com/register
  • Rotating knob for adjusting the speed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Digital Disperser Package
220/240 V
50 Hz
1 mL to 1.5 L
450 mm
LED
T 18 ULTRA-TURRAX™
5°C to 40°
3000 to 25,000 rpm
220/240 V, 50/60 Hz, 500 W
500 W
106 mm
87 mm
S 18 N - 19 G Dispersing tool
1 Ea.
5000 mPas
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ ULTRA-TURRAX™ T 18 Digital Disperser >