Learn More
SI Analytics™ Ubbelohde Viscometer (ASTM)
Ubbelohde viscometers are calibrated for automatic measurements
Brand: SI Analytics™ 285401255
Description
- Calibrated for automatic measurement
- ASTM
- Type No. 527 00
- Capillary No. 0
- Constant K = 0.001
Specifications
Ubbelohde Viscometer |