missing translation for 'onlineSavingsMsg'
Learn More
(+)-U-50488 hydrochloride, Tocris Bioscience™ Product Code.: 18045462

(+)-U-50488 hydrochloride, Tocris Bioscience™

Product Code. 18045462
10 mg
Click to view available options
Quantity:
10 mg
25 mg
50 mg
This item is not returnable. View return policy

Product Code. 18045462

Brand: Tocris Bioscience 0471/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective κ agonist

Less active enantiomer of (±)-U-50488 .

Racemate and (-)-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67197-96-0
Molecular Formula C19H26Cl2N2O·HCl
Synonym +/-u-50488 hydrochloride, +-u-50488 hydrochloride, +-trans-1r,2r-u-50488 hydrochloride, u-50488 hydrochloride, u50488 hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1r,2r-2-pyrrolidin-1-yl cyclohexyl acetamide hydrochloride, c19h26cl2n2o.hcl, +-trans-1r,2r0-u-50488 hcl, trans-2-3,4-dichlorophenyl-n-methyl-n-2-pyrrolidin-1-yl cyclohexyl acetamide hclShow MoreShow Less

Specifications

Quantity 10 mg
Formula Weight 405.79
Percent Purity >99%
Chemical Name or Material (+)-U-50488 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-U-50488 hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.