Learn More
Fisherbrand™ Tyvek™ Cat III Coverall
Shop All Fisherbrand Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Specifications
Specifications
Product Type | Hooded Coveralls |
Size | 2X-Large |
Material | Polyethylene |
Color | White |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Closure Type | Zipper |
Show More |
Documents
Scientific Resources
Flyers
Fisherbrand Tyvek Cat III Coveralls
File size: 230.5 kB
Fisherbrand Tyvek Cat III Coveralls
File size: 230.6 kB
Brochures
14177 FB NPI Fisherbrand Tyvek Cat III Coveralls EN
File size: 230.6 kB
Promotions
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Fisherbrand™ Tyvek™ Cat III Coverall >