missing translation for 'onlineSavingsMsg'
Learn More
Tyrphostin A9, 98+%, Thermo Scientific Chemicals Product Code.: 15425329

Tyrphostin A9, 98+%, Thermo Scientific Chemicals

Product Code. 15425329
100 mg
Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 15425329

Brand: Thermo Scientific Alfa Aesar J63058.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of PDGF receptor tyrosine kinase

Selective inhibitor of PDGF receptor tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10537-47-0
Molecular Formula C18H22N2O
Molecular Weight (g/mol) 282.387
MDL Number MFCD00209853
InChI Key MZOPWQKISXCCTP-UHFFFAOYSA-N
Synonym tyrphostin a9, malonoben, tyrphostin 9, malonoben ansi, caswell no. 291aa, 2-3,5-di-tert-butyl-4-hydroxybenzylidene malononitrile, unii-87te8mrs65, tyrphostin 9/ag-9, 3,5-di-t-butyl-4-hydroxybenzylidene malononitrile, epa pesticide chemical code 537200Show MoreShow Less
PubChem CID 5614
ChEBI CHEBI:82168
IUPAC Name 2-[(3,5-ditert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C=C(C#N)C#N

Specifications

Color Yellow
Quantity 100 mg
UN Number UN2811
Formula Weight 282.39
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tyrphostin A9

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY17H027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ63058.MF,J63058.MC
Lot NumberX04C025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ63058.MF,J63058.MC
Lot NumberF03X021 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ63058.MF,J63058.MC
Lot NumberF03X022 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ63058.MF,J63058.MC
Lot NumberU22D013 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ63058.MF,J63058.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tyrphostin A9, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.