Learn More
Thermo Scientific™ TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
Brand:  Thermo Scientific™ 75003683
441.88 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10586394
  • 512.55€ / Pack of 4
Edge
Explore more special offers
Description
Description
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
Features:
  • Adapters enable rotor to accept bottles up to 250 mL and tubes up to 100 mL
  • Maximum adapter capacity: 4 × 250 mL (4 × 400 mL with 400 mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125 mm (80 × 125 mm with 400 mL bio-bottles)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
50 mL Conical Tube
116 mm
4
Conical
29.5 mm
50 mL
Promotions
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Days
    4
    Hours
    11
    Minutes
    40
    Seconds
    25
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
    Days
    79
    Hours
    11
    Minutes
    40
    Seconds
    25
Product Content Correction
Product Title
Thermo Scientific™ TX-400 Rotor Adapters > 50mL Conical