Learn More
Tungsten Standard Metal Solution 1000ppm, for AAS, Solution NH3 0.1M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Physical Form | Liquid |
Color | Colorless |
Safety and Handling
Product Identifier
- Tungsten solution 1000 ppm
Product Content Correction
Product Title
Tungsten Standard Metal Solution 1000ppm, for AAS, Solution NH3 0.1M, Fisher Chemical™ >