Learn More
Agilent BioTek Tubing for 4 L Waste Bottle
For use with microplate washer systems
Brand:  Agilent BioTek 1263017
94.05 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 16502122
  • 99.00€ / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Description
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing
For use with microplate washer systems, For 4 L wash bottle
Product Title
Agilent BioTek Tubing for 4 L Waste Bottle > Tubing for 4 L Waste Bottle