missing translation for 'onlineSavingsMsg'
Learn More
Trimethylsulfonium iodide, 98%, Thermo Scientific Chemicals Product Code.: 11430483

Trimethylsulfonium iodide, 98%, Thermo Scientific Chemicals

Product Code. 11430483
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11430483

Brand: Thermo Scientific Alfa Aesar A12639.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2181-42-2
Molecular Formula C3H9IS
Molecular Weight (g/mol) 204.069
MDL Number MFCD00011632
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
Synonym trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340Show MoreShow Less
PubChem CID 75127
IUPAC Name trimethylsulfanium;iodide
SMILES C[S+](C)C.[I-]

Specifications

Melting Point ∼215°C (sublimation)
Quantity 1000 g
Beilstein 3555192
Sensitivity Light sensitive
Formula Weight 204.07
Percent Purity 99%
Chemical Name or Material Trimethylsulfonium iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241242 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA12639.30,A12639.0B,A12639.18
Lot Number10237693 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12639.30,A12639.0B,A12639.18
Lot Number10188315 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12639.30,A12639.0B,A12639.18
Lot Number10173327 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12639.30,A12639.0B,A12639.18
Lot Number10186874 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12639.30,A12639.0B,A12639.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethylsulfonium iodide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.