missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns

Use for laboratory-scale separations and intermediate purification based on different sizes of proteins, peptides, and other biomolecules. Cytiva Tricorn™ Superdex™ Prepacked Gel Filtration Columns are perfect for high-resolution applications.

2018.00€ - 2222.00€

Specifications

Buffer 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
Quantity Each
Includes Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
Material Borosilicate Glass
Particle Size 13μm
View More Specs

Includes: Valco fittings for AKTAdesign systems and are supplied with M6 connectors for FPLC System.

Products 4
Product Code Brand Product Type Bed Volume Price Quantity & Availability  
Product Code Brand Product Type Bed Volume Price Quantity & Availability  
10527280
View Documents
Cytiva
17-5174-01
Superdex 75 Prepacked Column 24 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
10362730
View Documents
Cytiva
17517501
Superdex 200 Prepacked Column 24 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
11535794
View Documents
Cytiva
28-9065-61
Superdex 200 Prepacked Column 3 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
11300292
View Documents
Cytiva
28-9205-04
Superdex 75 Prepacked Column 3 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

  • Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
  • Spherical composite of cross-linked agarose and dextran matrix
  • Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
  • Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200

Superdex Peptide:

  • Bed dimension: 10 x 300mm
  • Bed volume: 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 100 to 7000 (peptides)
  • pH Stability working range: 1 to 14
  • Column I.D.: 10mm
  • Bed height with one adaptor:     279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
Superdex 200:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
  • Pressure max.: 15 bar (217psi)
  • Separation range: 10,000 to 600,000 (globular peptides)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10mm
  • Bed height with one adaptor:     135 to 159mm (3mL column)
  • Bed height with two adaptors: 108 to 156mm (3mL column)
Superdex 75:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 3000 up to 70,000 (globular proteins)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10 mm
  • Bed height with one adaptor: 108 to 156mm (3mL column)
  • Bed height with two adaptors: 135 to 159mm (3mL column)

Specifications

Specifications

1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
13μm
4° to 30°C
High-resolution, semipreparative and analytical separations of biomolecules
Each
Borosilicate Glass
Spherical composite of cross-linked Agarose and Dextran
Prepacked Gel Filtration Column
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications