Learn More
Fisherbrand™ Translucent Silicone Tubing
Providing high tear strength tubing for the laboratory. Fisherbrand™ Translucent Silicone Tubing is available in a comprehensive range of sizes.
Brand:  Fisherbrand™ 1175-6599
 View more versions of this product
Product Code. 11756599
  • 182.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Autoclavable
  • Non-yellowing
  • High gloss finish
  • Excellent quality
  • Extended life in Peristaltic pumps
  • Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Specifications
FDA, BFR and WRAS approved
8 mm
12 mm
Autoclavable
Silicone
4 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Translucent Silicone Tubing > I.D.: 8mm; O.D.: 12mm; Length: 4m