Learn More
Fisherbrand™ Translucent Platinum-Cured Silicone Tubing
Use this tubing for pharmaceutical, biotech, peristaltic pumping and food applications. Fisherbrand™ Translucent Platinum-Cured Silicone Tubing is odorless, nontoxic and features excellent heat resistance.
Brand:  Fisherbrand™ 1176-8108
 View more versions of this product
Product Code. 11768108
  • 78.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Translucent
  • Non-yellowing
  • No peroxide by-products
  • Standard high tear strength
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • Extended life in Peristaltic pumps
Specifications
Specifications
FDA, BFR and WRAS approved
2.4 mm
4 mm
0.8 mm
-40°C
Translucent
200°C
Autoclavable
Platinum-cured Silicone
5 m
Biological , pharmaceutical, biotechnology, aerospace, hot work
Sterile
Flexible