missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Translucent Platinum Cured 5m Silicone Tubing
Providing a number of advantages over conventional peroxide cured silicones. Fisherbrand™ Translucent Platinum Cured 5m Silicone Tubing is available in a comprehensive range of stock sizes.
88.00€ - 1860.00€
Specifications
Certifications/Compliance | FDA, BFR and WRAS approved |
---|---|
Autoclavable | Autoclavable |
Overall Length (Metric) | 5 m |
Color | Translucent |
Flexibility | Flexible |
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Maximum Pressure | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Maximum Pressure | Price | Quantity & Availability | |||
11723974
|
Fisherbrand™
RESIL12519 |
12.5 mm | 5 m | 30 bar |
561.00€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
11778168
|
Fisherbrand™
1177-8168 |
12.8 mm | 5 m | 0.5 bar |
230.00€
Each |
In Stock
Log in to see stock. |
|||
11798168
|
Fisherbrand™
1179-8168 |
16 mm | 5 m | 0.5 bar |
279.00€
Each |
In Stock
Log in to see stock. |
|||
11798178
|
Fisherbrand™
1179-8178 |
19.2 mm | 5 m | 0.5 bar |
414.00€
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
11733974
|
Fisherbrand™
1173-3974 |
16 mm | 5 m | 30 bar |
1860.00€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
11773964
|
Fisherbrand™
1177-3964 |
3 mm | 5 m | 60 bar |
322.00€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
11728118
|
Fisherbrand™
1172-8118 |
4 mm | 5 m | 0.5 bar |
88.00€
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
11748138
|
Fisherbrand™
1174-8138 |
6.4 mm | 5 m | 1 bar |
176.00€
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
11783964
|
Fisherbrand™
1178-3964 |
5 mm | 5 m | 40 bar |
357.00€
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||
11788128
|
Fisherbrand™
1178-8128 |
6.4 mm | 5 m | 0.5 bar |
110.00€
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||
11793964
|
Fisherbrand™
RESIL6119 |
6 mm | 5 m | 40 bar |
394.00€
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||
11758148
|
Fisherbrand™
1175-8148 |
8 mm | 5 m | 0.5 bar |
170.00€
Each |
In Stock
Log in to see stock. |
|||
11703974
|
Fisherbrand™
1170-3974 |
8 mm | 5 m | 35 bar |
592.00€
Each |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||
11738158
|
Fisherbrand™
1173-8158 |
9.6 mm | 5 m | 1 bar |
206.00€
Each |
In Stock
Log in to see stock. |
|||
11713974
|
Fisherbrand™
RESIL95159 |
9.5 mm | 5 m | 30 bar |
583.00€
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||
11718158
|
Fisherbrand™
1171-8158 |
9.6 mm | 5 m | 0.5 bar |
178.00€
Each |
In Stock
Log in to see stock. |
|||
11738168
|
Fisherbrand™
1173-8168 |
9.6 mm | 5 m | 1 bar |
251.00€
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||
11798148
|
Fisherbrand™
1179-8148 |
8 mm | 5 m | 1.5 bar |
203.00€
Each |
In Stock
Log in to see stock. |
|||
Description
- Autoclavable
- Non-yellowing
- High gloss finish
- Excellent quality
- Highly hydrophobic
- No peroxide byproducts
- High tear strength as standard
- Extended life in Peristaltic pumps
- Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
FDA, BFR and WRAS approved | |
5 m | |
Flexible |
Autoclavable | |
Translucent |