Learn More
Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Providing a number of advantages over conventional peroxide cured silicones. Fisherbrand™ Translucent Platinum Cured Silicone Tubing is available in a comprehensive range of stock sizes.
Brand:  Fisherbrand™ 1172-8138
 View more versions of this product
Product Code. 11728138
  • 136.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Autoclavable
  • Non-yellowing
  • High gloss finish
  • Excellent quality
  • Highly hydrophobic
  • No peroxide byproducts
  • High tear strength as standard
  • Extended life in Peristaltic pumps
  • Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
Specifications
FDA, BFR and WRAS approved
11.2 mm
1bar
6.4 mm
5 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Translucent Platinum Cured Silicone Tubing > I.D.: 6.4mm; O.D.: 11.2mm; Length: 5m; Nominal Pressure: 1bar