Learn More
trans-beta-Styrylboronic acid, 97%, Thermo Scientific Chemicals
Description
trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Specifications
Specifications
Melting Point | 98°C to 100°C |
Molecular Formula | C11H10N4O4 |
Quantity | 1 g |
Synonym | carbadox, unii-m2x04r2e2y, getroxel, mecadox, dsstox_cid_23913, dsstox_rid_80088, dsstox_gsid_43913, fortigro, karbadox czech, methyl n-e-1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene methyl iminocarbamate |
InChI Key | BPMVRAQIQQEBLN-UHFFFAOYSA-N |
SMILES | COC(=O)N=NC=C1C=[N+](C2=CC=CC=C2N1O)[O-] |
IUPAC Name | methyl N-[(E)-(1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene)methyl]iminocarbamate |
Molecular Weight (g/mol) | 262.23 |
PubChem CID | 5353472 |
Formula Weight | 147.97 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
5003Q11W | Certificate of Analysis | 26/12/2024 | H53227.06,H53227.03 |
5007R23T | Certificate of Analysis | 07/12/2021 | H53227.06,H53227.03 |
5011D15M | Certificate of Analysis | 07/12/2021 | H53227.06,H53227.03 |
5019D14Q | Certificate of Analysis | 07/12/2021 | H53227.06,H53227.03 |
5004K01Q | Certificate of Analysis | 07/12/2021 | H53227.06,H53227.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.