Learn More
Thermo Scientific™ TracePLOT™ TG-BOND Alumina GC Columns: KCl Deactivation
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
30 m
50 m
Diameter (Metric):
0.32 mm
0.53 mm
Unit Size:
Each
Beschreibung
Use Thermo Scientific™ TracePLOT TG-BOND Alumina GC Columns for linear and quantitative analysis of polar unsaturated hydrocarbons.
Applications:
- C1-C5 hydrocarbons
- Unsaturated hydrocarbon isomers
- Strong bonding prevents damage to injection and detection systems from particle release during valve-switching operations.
- Columns may be regenerated to restore full efficiency and selectivity after water adsorption.
- Rigorous testing ensures proper film thickness (1,3-butadiene), selectivity (propadiene, methyl acetylene), resolution (trans-2-butene, 1-butene) and coating efficiency (1,3-butadiene).
- Alumina-PLOT
- AT™-Alumina
- CP-Al2O3/KCl
- CP-Al2O3/Na2SO4
- GS-Alumina
- GS-Alumina KCl
- HP PLOT M
- HP PLOT S
- Rt™-Alumina BOND (KCl)
- Rt™-Alumina BOND (Na2SO4 )
Spezifikation
Spezifikation
| Max. Temperature | Up to 200°C |
| Diameter (Metric) | 0.53 mm |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | TracePLOT™ TG-BOND Alumina |
| For Use With (Application) | Polar Unsaturated Hydrocarbons Analysis, GC for Unsaturated Hydrocarbon Isomers, Linear and Quantitative Analysis of Polar Unsaturated Hydrocarbons |
| Film Thickness | 10 μm |
| Length (Metric) | 30 m |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur