missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Total Reactive Oxygen Species (ROS) Assay Kit 520 nm

Contains the necessary regent and buffer for identifying ROS in cells by flow cytometry in the FITC channel. Reactive oxygen species are chemically reactive oxygen-containing molecules that are generated as a natural byproduct of the oxygen metabolism.

Brand:  Invitrogen™ 88-5930-74

250.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 15580977

  • 376.00€ / 200 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes:
  • ROS Assay Stain Concentrate (lyophilzed powder): 1 vial
  • ROS Assay Buffer
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 tests
200 tests
495/520 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE143109 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number88-5930-74
Lot NumberE143264 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number88-5930-74
Lot NumberE143225 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number88-5930-74
Lot NumberE141274 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number88-5930-74
Lot NumberE141474 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number88-5930-74
Promotions

Promotions

  • Multiple Manufacturers
    Now extended - offer ends 28 March
    Promo Code: 24111

    Expires2025-03-28

    Days

    0

    Hours

    21

    Minutes

    59

    Seconds

    14
    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Total Reactive Oxygen Species (ROS) Assay Kit 520 nm > Total Reactive Oxygen Species (ROS) Assay Kit, 520nm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.