Learn More
Total Organic Carbon Standard, Reagecon™ Product Code.: 12086546
Total Organic Carbon Standard, Reagecon™
BIOPHARMA
Product Code. 12086546 Shop All Reagecon Products
500 mL
Click to view available options
Quantity:
500 mL
Product Code. 12086546
Brand: Reagecon TOC1M
Specifications
Quantity 500 mL
Concentration 1000 ppm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberTOC1M24J1 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberTOC1M
Lot NumberTOC1M24E1 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberTOC1M
Lot NumberTOC1M24B1 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog NumberTOC1M
Lot NumberTOC1M23L1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberTOC1M
Lot NumberTOC1M23H1 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberTOC1M
Product Content Correction
Product Title
Total Organic Carbon Standard, Reagecon™ >