Learn More
Toluene ≥ 99.5%, BAKER ANALYZED™ ACS, J.T. Baker™ Product Code.: 10741542
Toluene ≥ 99.5%, BAKER ANALYZED™ ACS, J.T. Baker™
Product Code. 10741542
2.5 L
Click to view available options
Quantity:
2.5 L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10741542
Brand: JT Baker 8077.2500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Exceeds ACS Specifications
Chemical Identifiers
CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
IUPAC Name toluene
SMILES CC1=CC=CC=C1
Specifications
Melting Point -95°C
Color Max. 10 (APHA)
Density 0.867 g/cm³ (20°C)
Boiling Point 110.6°C (1013 hPa)
Quantity 2.5 L
Flash Point 4°C
Assay Percent Range min. 99.5%
Residue after Evaporation max. 0.001%
Water max. 0.03%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction
Product Title
Toluene ≥ 99.5%, BAKER ANALYZED™ ACS, J.T. Baker™ >