missing translation for 'onlineSavingsMsg'
Learn More
TMPyP4 tosylate, Tocris Bioscience™ Product Code.: 18074662

TMPyP4 tosylate, Tocris Bioscience™

Product Code. 18074662
50 mg
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy

Product Code. 18074662

Brand: Tocris Bioscience 4253/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of human telomerase

Cationic porphyrin; inhibits human telomerase. Stacks with G tetrads to stabilize quadruplex DNA. Inhibits cell proliferation and induces cell death in three myeloma cell lines. Activity results in telomere shortening at concentrations between 1 and 5 μM.
TRUSTED_SUSTAINABILITY

Specifications

CAS 36951-72-1
Molecular Formula C72H66N8O12S4
Quantity 50 mg
Synonym tmpyp, tmpyp4 tosylate, tmpyp tetratosylate, tmpyp4 tetratosylate, tppt 4, meso-tetra 4-n-methyl-pyridyl porphyrin, meso-tetrakis n-methyl-4-pyridyl porphine tetratosylate salt, meso-tetrakis n-methylpyridinium-4-yl porphine tetratosylate, meso-tetrakis n-methyl-4-pyridyl porphine tetrakis p-toluenesulfonate, 5,10,15,20-tetrakis 1-methylpyridinium-4-yl porphyrin tetra p-toluenesulfonate
InChI Key AKZFRMNXBLFDNN-UHFFFAOYSA-K
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=[N+](C=C7)C)C8=CC=[N+](C=C8)C)C9=CC=[N+](C=C9)C)N3
IUPAC Name 4-methylbenzenesulfonate;5,10,15,20-tetrakis(1-methylpyridin-1-ium-4-yl)-21,22-dihydroporphyrin
Molecular Weight (g/mol) 1363.604
PubChem CID 11979833
Formula Weight 1363.6
Percent Purity >95%
Chemical Name or Material TMPyP4 tosylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TMPyP4 tosylate, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.