missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TLR4 Inhibitor Peptide Set

Applications: Flow Cytometry, Functional, In vitro assay, Binding Inhibition, Block/Neutralize

Brand:  Novus Biologicals™ NBP2-26244

Product Code. 18710534

  • 579.00€ / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Consists of 11 amino acid (KYSFKLILAEY) inhibitory sequence, derived from A46 vaccinia virus protein, linked to 9R (RRRRRRRRR) homopolymer delivery sequence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
2 Vials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ TLR4 Inhibitor Peptide Set > TLR4 Inhibitor Peptide Set

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only