Learn More
Lovibond™ Tintometer™ SensoDirect 150 All-In-One Handheld Meter, pH/Redox
Features include automatic calibration, buffer recognition, large LCD display and result hold function. Lovibond SensoDirect 150 is combined instrument capable of measuring pH, ORP, Conductivity, TDS, Dissolved Oxygen and Temperature in a single meter.
Brand:  Lovibond™ 724230
Product Code. 12631745
  • 769.00€ / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes:
  • Instrument
  • Batteries
  • pH electrode
  • Temperature probe
  • Redox electrode
  • pH buffer set with pH 4.0 and pH 7
  • Instruction manual
  • Guarantee sheet in case
  • Features
  • pH/Redox, Temperature °C/°F
  • All in one
  • Real time data logger
  • Protective casing
  • RS 232 / USB
Specifications
Specifications
0 to 14 pH, ±0.02 pH + 2 digits
±0.8°C (1.5°F)
220 x 120 x 40 mm
0 to 14
mV to 1999mV to 1999mV
(Range/Resolution) 0 to 200.0µS / 0.1µS; 0.2 to 2.000mS / 0.001mS; 2 to 20.00mS / 0.01mS; 20 to 200.00mS / 0.1mS
0 to 14pH, 0.01 pH
Manual: 0°C to 100°C, Automatic (ATC)
625 g
0 to 1999mV, 1mV
SensoDirect 150 Set pH/Redox
Measures pH/Redox, dissolved oxygen and conductivity/TDS
0 to 200.0 µS
0 to 132,000ppm
0 to 1999mV, 1mV
0.1°C
0°C to 50°C
CE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lovibond™ Tintometer™ SensoDirect 150 All-In-One Handheld Meter, pH/Redox > SensoDirect 150 All-In-One Handheld Meter, pH/Redox