Learn More
Fisherbrand™ Three-Sided Front Gamma Radiation Shield
Provides radiation protection shielding for use in laboratories.
Brand: Fisherbrand™ CSR-S3G
Description
- Transparent shielding
- Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Acrylic | |
Three sides front, Sides | |
Front: L x W: 50 x 46 cm (19.68 x 18.11 in.), Sides: 50 x 30 cm (19.68 x 11.81 in.) |
Gamma radiation protection | |
Radiation shielded box |
Product Suggestions
Customers who viewed this item also viewed.