Learn More
Codan™ Three-part Syringes
Use Codan™ Three-part Syringes to accommodate a variety of medical needs including: catheterization, hypodermic ear treatments, and more. The syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spill
Brand:  Codan™ 62.2611
 View more versions of this product
Product Code. 11348763
Description
Description
  • Sterile
  • Graduations
  • Disposable
  • Latex-free
Specifications
Specifications
Autoclavable
Dual scale; Silicone seal
Single-use
2 mL
Yes
Polypropylene
Hypodermic
0.1 mL
Centric Luer Cone
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Codan™ Three-part Syringes > Connection Type: Luer centered cone; Volume: 2mL