Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
Brand:  Greiner Bio-One 662630
 View more versions of this product
Product Code. 10733544
  • 284.83€ / Pack of 48
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
TRUSTED_SUSTAINABILITY
Specifications
Specifications
33.6 mm2
0.1 to 0.35 mL
3.0 μm
24
Polystyrene
0.6 x 106 pores/cm2
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Greiner Bio-One ThinCert™ Tissue Culture Inserts > Format: 24 well; Pore size: 3.0μm; Pore Density: 0.6 x 106 sq. cm