Learn More
Brady™ Thermal Transfer Printer Labels: Rectangular
Rectangular printable labels for thermal transfer printers.
166.00€ - 435.00€
Specifications
Color White
Quantity 2000/Roll
View More Specs
Products 17
Product Code Brand Product Type Quantity Height (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Product Type Quantity Height (Metric) Width (Metric) Price Quantity & Availability  
17110161
View Documents
Brady™
121006
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 6.35 mm
264.00€
Pack of 2000
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17120161
View Documents
Brady™
121007
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 12.70 mm
230.00€
Pack of 2000
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17100161
View Documents
Brady™
121005
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 33.0 mm
216.00€
Pack of 2000
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
17170161
View Documents
Brady™
121013
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 19.05 mm
352.00€
Pack of 2000
Estimated Shipment: 23-04-2025
to see stock.
Add to basket
17140161
View Documents
Brady™
121010
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 25.4 mm
236.00€
Pack of 2000
In Stock
to see stock.
Add to basket
17150151
View Documents
Brady™
121000
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 9.525 mm
166.00€
Pack of 2000
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17140151
View Documents
Brady™
120999
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 12.70 mm
181.00€
Pack of 2000
In Stock
to see stock.
Add to basket
17180151
View Documents
Brady™
121003
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 19.05 mm
334.00€
Pack of 2000
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
17190151
View Documents
Brady™
121004
Thermal Transfer Printer Labels - 3 in. Core 2000/Roll 25.4 mm
169.00€
Pack of 2000
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
17130161
View Documents
Brady™
121009
Thermal Transfer Printer Labels - 3 in. Core 12.70 mm
236.00€
Pack of 2000
In Stock
to see stock.
Add to basket
17160161
View Documents
Brady™
121012
Thermal Transfer Printer Labels - 3 in. Core 44.45 mm
379.00€
Pack of 2000
In Stock
to see stock.
Add to basket
17150161
View Documents
Brady™
121011
Thermal Transfer Printer Labels - 3 in. Core 25.4 mm
180.00€
Pack of 2000
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
17180161
View Documents
Brady™
121018
Thermal Transfer Printer Labels - 1 in. Core 9.525 mm
191.00€
Pack of 2000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
17110171
View Documents
Brady™
121021
Thermal Transfer Printer Labels - 1 in. Core 2000/Roll 19.05 mm
435.00€
Pack of 2000
In Stock
to see stock.
Add to basket
17120171
View Documents
Brady™
121028
Thermal Transfer Printer Labels - 1 in. Core 2000/Roll 25.4 mm
272.00€
Pack of 2000
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17140171
View Documents
Brady™
121031
Thermal Transfer Printer Labels - 1 in. Core 19.05 mm 25.40 mm
404.00€
Pack of 2000
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17130171
View Documents
Brady™
121029
Thermal Transfer Printer Labels - 1 in. Core 2000/Roll 25.4 mm
208.00€
Pack of 2000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
Description
Description
Printer Compatibility
  • BBP81
  • Brady 1244
  • Brady 1344
  • Brady 200MVP Plus
  • Brady 2461
  • Brady 300MVP Plus
  • Brady 300X-Plus II
  • Brady 3481
  • Brady 360X-Plus II
  • Brady 600X-Plus II
  • Brady 6441
  • Brady IP
  • Tagus T200
  • Tagus T300
  • Thermal Transfer Printers
This material is UL recognized with its respective ribbon; use Brady Ribbon Part# R4902.
Order Info
See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications
Specifications
White
2000/Roll
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Brady™ Thermal Transfer Printer Labels: Rectangular