missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TH 9619 disodium salt

MTHFD2 inhibitor

178.00€ - 626.00€

Specifications

Chemical Name or Material Disodium N-[[5-[[2-(2,4-Diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)acetyl]amino]-3-fluoro-2-pyridinyl]carbonyl]-L-glutamate
Target Dehydrogenase Inhibitors
Molecular Formula C17H16FN7Na2O7
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18055112
View Documents
Tocris Bioscience™
7719/50
50 mg
626.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
18014022
View Documents
Tocris Bioscience™
7719/10
10 mg
178.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TH 9619 disodium salt is a potent and selective methylenetetrahydrofolate dehydrogenase inhibitor (MTHFD2; IC50 = 47 nM). It shows efficacity on HL-60 cell viability with EC50 of 11nM. In vitro, It shows antiproliferative efficacy in acute myeloid leukemia (AmL) cells and T-ALL Jurkat cells. In vivo, TH9619 disodium salt impairs cancer progression in a mouse disease model of AmL.
Specifications

Specifications

Disodium N-[[5-[[2-(2,4-Diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)acetyl]amino]-3-fluoro-2-pyridinyl]carbonyl]-L-glutamate
Dehydrogenase Inhibitors
C17H16FN7Na2O7
0.98
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.