Learn More
WTW™ TetraCon™ 925 4 Electrode Conductivity Cells
With its easy handling and robust design, these electrode conductivity cells are proven 2 and 4 electrode technology. The four electrode measuring cell TetraCon™ 925 model is for medium and high conductivity.
Brand: WTW™ 301710
620.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Graphite four electrode measuring cell, 1.5 fixed cable
- IDS conductivity measuring cells – measurements from pure water all the way to concentrated solutions with proven two and four electrode technology
- Easiest handling, robust design
- Wide area of application - from pure water to highly concentrated solutions
- The four electrode measuring cell TetraCon™ 925 model is for medium and high conductivity
Specifications
4-electrodes, Graphite | |
0°C to 100°C | |
120 mm |
1.5 m | |
15.3 mm |
Product Content Correction
Product Title
WTW™ TetraCon™ 925 4 Electrode Conductivity Cells > Cell Constant: 0.475cm; Cable Length: 1.5m; Material: Epoxy